

Isopropyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-21-4
:
Formula:C₅H₁₀O₂
MDL Number:MFCD00008877
:102.13 g/mol
Merck Index:13,05225
Boiling Pt:85 °C (1013 hPa)
Melting Pt:–62 °C
Density:0.8732 g/cm³ (20 °C)
EINECS:203-561-1
UN:1220
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:4 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Solubility:31 g/l (20 °C)
- Melting Point:-73 °C
- Molar Mass:102.13 g/mol
- Boiling Point:88 - 90 °C (1013 hPa)
- Vapor Pressure:61 hPa (20 °C)
- Flash Point:1.8 - 8 %(V)
- Refractive Index:4 °C
- Density:0.87 g/cm3 (20 °C)
- Ignition Point:460 °C
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Related Information
Frequently Bought Together