Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Isopropyl acetate 99+%, pure
 
undefined
 
 
Isopropyl acetate 99+%, pure

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3760 to 1.3780 (20°C, 589 nm)
  • Water:
    ≤0.1 %
  • Density:
    (20°C) 0.870 to 0.874 kg/l
  • GC:
    ≥99.0 %
  • Residue after evaporation:
    ≤0.01 %

 

 

Frequently Bought Together