Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
    
 
undefined
 
 
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
    
 
 
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

 

 

  • Assay (on anhydrous substance):
    Min. 99,7 %
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0,2 %
  • Pesticide analysis (Lindane/ECD):
    Max. 5 ng/l

 

Frequently Bought Together