


Acetone ≥99.4% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
:Packaged under inert gas.
Flash Pt:<–20 °C (closed cup)
REACH:01-2119471330-49
:GHS02
:GHS07
:GHS08
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Min. 99.4 %
- Assay ((CH₃)₂CO) (by GC, corrected for water):≥99.4 %
- Color (APHA):≤10
- Residue after Evaporation:≤1.0 ppm
- Substances Reducing Permanganate:Passes Test
- Titrable Acid (µeq/g):≤0.3
- Titrable Base (µeq/g):≤0.6
- Water (H₂O):≤0.5 %
- FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤5
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤10
Specification Test Results
Frequently Bought Together