

Acetone 99.5% for electronic use (MOS) residue-free
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56 °C (760 mmHg)
Melting Pt:–95 °C
Density:0.79 g/cm³
Flash Pt:–20 °C
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.2 % (Coulometric)
- GC:≥99.4 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
- Acidity:≤0.0003 meq/g
- Chloride (Cl):≤2 ppm
- Copper (Cu):≤10 ppb
- Iron (Fe):≤50 ppb
- Methanol:≤0.05 % (GC)
Specification Test Results
Frequently Bought Together