

Methyl ethyl ketone ≥99.7%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:78-93-3
:
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648
Merck Index:13,06097
Boiling Pt:79.6 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.8054 g/cm³ (20 °C)
Flash Pt:–4 °C (DIN 51755 Part 1)
EINECS:201-159-0
UN:1193
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
:72.11 g/mol
REACH:01-2119457290-43
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.7 %
- Acidity (at filling):Max. 0.0005 meq/g
- Evaporation residue:Max. 5 ppm
- Water:Max. 0.1 %
- Transmittance (335 nm):Min. 40 %
- Transmittance (340 nm):Min. 80 %
- Transmittance (350 nm):Min. 98 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
spec_VWRC25643.294 spec_VWRC25643.294 spec_VWRC25643.328 spec_VWRC25643.328
Product specification
Frequently Bought Together