

Methyl ethyl ketone ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:78-93-3
:
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648
Merck Index:13,06097
Boiling Pt:79…81 °C
Melting Pt:–87 °C
Density:0.805 g/cm³ (20 °C)
Flash Pt:11.5 %(V)
Storage Temperature:Ambient
UN:1193
ADR:3,II
:72.11 g/mol
EINECS:201-159-0
REACH:01-2119457290-43
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Assay (GC):Min. 99.5 %
- Boiling range:78.5 - 80.5 °C
- Autoignition Temperature:960 °F
- Density (D 20/4):0.804 - 0.806
- Refractive index (n 20/D):1.3770 - 1.3810
- Non-volatile matter:Max. 0.002 %
- Water (Karl Fischer):Max. 0.05 %
- Titrable Acid:Max. 0.0005 meq/g
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Zinc (Zn):Max. 0.00002 %
- 2-Butanol (GC):Max. 0.05 %
- KMnO₄ reducing matter (as O):Max. 0.0003 %
- APHA:Max. 15
- Explosion Limit:10.1 %
Specification Test Results
Frequently Bought Together