

Methyl ethyl ketone, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
: Avantor
:78-93-3
:
:C₂H₅COCH₃
:MFCD00011648
:13,06097
:79.6 °C (1013 hPa)
:–86 °C
:0.8054 g/cm³ (20 °C)
:–4 °C
:Ambient
:1193
:3,II
:72.11 g/mol
:201-159-0
:01-2119457290-43
:GHS02
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Assay (by GC):min. 99 %
- Acidity (as CH3COOH):max. 0.003 %
- Boiling Range:max. 1.5 °C
- Density (g/ml) at 25°C:0.797 - 801.000
- Recorded Boiling Point:79.6 °C
- Residue after Evaporation:max. 0.005 %
- Water (H2O):max. 0.2 %
- Aluminium (Al):max. 0.5
- Boron (B):max. 0.02
- Barium (Ba):max. 0.1
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.05
- Cobalt (Co):max. 0.02
- Chromium (Cr):max. 0.02
- Copper (Cu):max. 0.02
- Iron (Fe):max. 0.1
- Magnesium (Mg):max. 0.1
- Manganese (Mn):max. 0.02
- Nickel (Ni):max. 0.02
- Lead (Pb):max. 0.1
- Tin (Sn):max. 0.1
- Zinc (Zn):max. 0.1
Frequently Bought Together