Orders LinkContactUsLinkComponent
Stopcocks, for storage tanks, Nalgene®
Stopcocks, for storage tanks, Nalgene®
   Sustainable Options Available
Stopcocks, for storage tanks, Nalgene®
   Sustainable Options Available
Certificates
Stopcocks, for storage tanks, Nalgene®
2 Options Available Below
Product Details & Documents
LDPE, white.
  • Good chemical resistance
  • Godd temperature resistance
Ensure safe and efficient liquid dispensing from tanks by using Nalgene™ spigots for storage tanks. Spigots are for use with Naglene™ tanks up to 100 gal. with factory installed threaded boss.
Specifications for Product Family
Specifications
  • Catalog No:
  • NALG6420-0500
  • NALG6420-0750
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Thread
    Availability
    Price
  • 12,7 mm NPT, male thread
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 61.60 SFr.
    61.60 SFr.
    1 * 1 ST
       61.60 SFr.
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG6420-0500
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 19 mm NPT, male screw thread
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 67.20 SFr.
    67.20 SFr.
    1 * 1 ST
       67.20 SFr.
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG6420-0750
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact