Order Entry
Switzerland
ContactUsLinkComponent
2-Pentanone 99%, pure
2-Pentanone 99%, pure
Certificates
undefined
2-Pentanone 99%, pure

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3890 to 1.3910 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together