Order Entry
Switzerland
ContactUsLinkComponent
2-Octanone
2-Octanone
Catalog # ROTH9440.1
Supplier:  Roth Carl
CAS Number:  
2-Octanone
Catalog # ROTH9440.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :128.21 g/mol
Merck Index:13,04731
Boiling Pt:173 °C (1013 hPa)
Melting Pt:–16 °C
Density:0.8171 g/cm³ (20 °C)
Flash Pt:56 °C
Storage Temperature:Ambient
EINECS:203-837-1
UN:1993
ADR:3,III
CAS Number:111-13-7
Formula:C₈H₁₆O
MDL Number:MFCD00009540