

2-Octanone 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-13-7
:
Formula:C₈H₁₆O
MDL Number:MFCD00009540
:128.21 g/mol
Merck Index:13,04731
Boiling Pt:173 °C (760 mmHg)
Melting Pt:–16 °C
Density:0.819 g/cm³
Flash Pt:55 °C
Storage Temperature:Ambient
EINECS:203-837-1
UN:1224
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4140 to 1.4160 (20°C, 589 nm)
- GC:≥99.0 %
- Clarity of solution:Passes test
Specification Test Results
Frequently Bought Together