N,O-Bis(trimethylsilyl)trifluoroacetamide with 10% trimethylchlorosilane
Catalog # HYPETS-38840
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 Ampoul
- Seq:0510
Specifications
About this item
:
:257.4 g/mol
EINECS:247-103-9
CAS Number:25561-30-2
Formula:C₈H₁₈F₃NOSi₂
MDL Number:MFCD00008269