Order Entry
Switzerland
ContactUsLinkComponent
2-Heptanone
2-Heptanone
Catalog # EHERC14127600
Supplier:  EHRENSTORFER
CAS Number:  
2-Heptanone
Catalog # EHERC14127600
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070
  • Environmentally Preferable:

Specifications

About this item

 :
 :114.19 g/mol
Merck Index:13,04683
Boiling Pt:150 °C (1013 hPa)
Melting Pt:–35 °C
Density:0.8111 g/cm³ (20 °C)
Flash Pt:43 °C
Storage Temperature:Ambient
EINECS:203-767-1
UN:1110
ADR:3,III
 :GHS02
 :GHS07
CAS Number:110-43-0
Formula:C₇H₁₄O
MDL Number:MFCD00009513