
Silver sulphate 97+%
Catalog # A16324.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:311.8 g/mol
Merck Index:13,08603
Boiling Pt:1085 °C (1013 hPa)
Melting Pt:655 °C
Density:5.45 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-653-7
:GHS05
CAS Number:10294-26-5
Formula:Ag₂SO₄
MDL Number:MFCD00003407