Pronase E (from Streptomyces griseus)
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
Specifications
- CAS No.:9036-06-0
- Source:Streptomyces griseus
- Storage conditions:Freezer
- MDL No.:MFCD02253017
- Enzyme name:Pronase E
Specifications
Frequently Bought Together