Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Proteinase K
Proteinase K
Proteinase K
Catalog # PIER17916
Supplier:
Thermo Fisher Scientific
CAS Number:
Expand image
Proteinase K
Catalog # PIER17916
Supplier:
Thermo Fisher Scientific
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
100 mg
CAS No.:
39450-01-6
Storage conditions:
Freezer
Enzyme name:
Proteinase K
Specifications
About this item
Product Details
Product Details