



Acrylic acid stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:79-10-7
:
Formula:C₃H₄O₂
MDL Number:MFCD00004367
:72.06 g/mol
Boiling Pt:141.6 °C (1013 hPa)
Melting Pt:13 °C
Density:1.06 g/cm³ (10 °C)
Storage Temperature:Ambient
EINECS:201-177-9
UN:2218
ADR:8 (3),II
:M-Clarity™ quality level = MQ200
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
:GHS02
:GHS05
:GHS07
:GHS09
Specification Test Results
- Solubility:1000 g/l (25 °C)
- Melting Point:13 °C
- Molar Mass:72.06 g/mol
- Boiling Point:141 °C (1013 hPa)
- Vapor Pressure:3.8 hPa (20 °C)
- Flash Point:46 °C
- Refractive Index:1.4224 (20 °C, 589 nm)
- Explosion Limit:3.9 - 19.8 %(V)
- Density:1.05 g/cm3 (20 °C)
- pH:2.1 (72.06 g/l, H2O, 20 °C)
- Ignition Point:390 °C DIN 51794
- Synonyms:2-Propenoic acid, Acroleic acid
- Hill Formula:C3H4O2
Specification Test Results
Frequently Bought Together