2-Decanone, Supelco®
Catalog # 68228-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
Boiling Pt:211 °C (1013 hPa)
Melting Pt:3 °C
Density:0.82 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:156.27 g/mol
Storage Temperature:Ambient
EINECS:211-752-6
CAS Number:693-54-9
Formula:C₁₀H₂₀O
MDL Number:MFCD00009571