Order Entry
Switzerland
ContactUsLinkComponent
Palladium(II) acetate (47.5% Pd)
Corrosive
Palladium(II) acetate (47.5% Pd)
Certificates
undefined
Corrosive
Palladium(II) acetate (47.5% Pd)

Specification Test Results

  • Appearance:
    Orange to brown Powder
  • Infrared spectrum:
    Conforms
  • Assay:
    46.9 to 48.5 % (Pd)
  • Additional info:
    Platinum group metals (Pt+Ru+Rh+Ir): 300 ppm
  • Chloride (Cl):
    ≤1000 ppm
  • Aluminium (Al):
    ≤50 ppm
  • Calcium (Ca):
    ≤50 ppm
  • Chromium (Cr):
    ≤10 ppm
  • Copper (Cu):
    ≤50 ppm
  • Iron (Fe):
    ≤50 ppm
  • Lead (Pb):
    ≤50 ppm
  • Magnesium (Mg):
    ≤50 ppm
  • Manganese (Mn):
    ≤10 ppm
  • Nitrogen (N):
    ≤2 %
  • Silicon (Si):
    ≤50 ppm
  • Zinc (Zn):
    ≤25 ppm

Specification Test Results

Related Information

Frequently Bought Together