Dihydrocytochalasin B, Calbiochem®
Catalog # 250225-1
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:481.63 g/mol
Melting Pt:198…203 °C
EINECS:254-324-4
CAS Number:39156-67-7
Formula:C₂₉H₃₉NO₅