Order Entry
Switzerland
ContactUsLinkComponent
Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 3001100
Irritant
Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 3001100
Catalog # REGE3001100
Supplier:  REAGECON
CAS Number:  
Irritant
Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 3001100
Catalog # REGE3001100
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :332.24 g/mol
Merck Index:13,02002
EINECS:237-029-5
UN:3264
ADR:8,III
 :GHS07
CAS Number:13590-82-4
Formula:Ce(SO₄)₂
MDL Number:MFCD00148852