SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Enzymes
Pineapple Bromelain (from Stem)
Pineapple Bromelain (from Stem)
MOLE36924643-25G
:
Molekula
:
Pineapple Bromelain (from Stem)
MOLE36924643-25G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
25 g
CAS No.:
37189-34-7
Source:
Stem
Species:
Pineapple
Environmentally Preferable:
Enzyme name:
Bromelain