Order Entry
Switzerland
ContactUsLinkComponent
Bromelain (from stem)
Bromelain (from stem)
Catalog # PROOFIP4
CAS Number:  
Bromelain (from stem)
Catalog # PROOFIP4
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • CAS No.:
    37189-34-7
  • Source:
    Stem
  • Enzyme name:
    Bromelain

Specifications

About this item