Bilirubin, Millipore®
Glass bottle for solids, 1 g
Supplier: Merck Millipore (Calbiochem)
Jump to:
Product Details & Documents
Synonyms: 3-(2-((3-(2-Carboxyethyl)-4-methyl-5-((Z)-(3-methyl-5-oxo-4-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-2-yl)methyl)-4-methyl-5-((Z)-(4-methyl-5-oxo-3-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-3-yl)propanoic acid
- CAS Number:635-65-4
- Formula:C₃₃H₃₆N₄O₆
- MDL Number:MFCD00005499
- Molecular Weight:584.67 g/mol
- Melting Pt:> 360 °C
- Density:1.31 g/cm³ (20 °C)
- EINECS:211-239-7
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



