Order Entry
Switzerland
ContactUsLinkComponent
4'-Isobutylacetophenone ≥96.0% (by GC)
4'-Isobutylacetophenone ≥96.0% (by GC)
Supplier:  TCI
Certificates
undefined
4'-Isobutylacetophenone ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Yellow to Green clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

Frequently Bought Together