Pesticide reference standard, 3-Chloroaniline 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA11351000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:108-42-9
- Description:3-Chloroaniline 100 µg/ml in methanol
- Formula:C₆H₆ClN
- Description:3-Chloroaniline