Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Hydrocortisone 98%

Hydrocortisone 98%

Health
Hydrocortisone 98%
Certificates
Hydrocortisone 98%

4 Options Available Below

Product Details & Documents
  • CAS Number:50-23-7
  • Formula:C₂₁H₃₀O₅
  • MDL Number:MFCD00011654
  • Molecular Weight:362.47 g/mol
  • Boiling Pt:567 °C (1013 hPa)
  • Melting Pt:212…217 °C
  • Density:1,4 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-020-1

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    212 to 217 °C
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +150° to +156° (20 °C, 589 nm) (C=1, dioxane)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 44.30 SFr.
    44.30 SFr.
    1 * 1 g
       44.30 SFr.

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 167.00 SFr.
    167.00 SFr.
    1 * 5 g
       167.00 SFr.

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 622.00 SFr.
    622.00 SFr.
    1 * 25 g
       622.00 SFr.

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1 870.00 SFr.
    1 870.00 SFr.
    1 * 100 g
       1 870.00 SFr.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.