Pesticide reference standard, 2,5-Dichlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12452000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:583-78-8
- Description:2,5-Dichlorophenol 100 µg/ml in methanol
- Formula:C₆H₄Cl₂O
- Description:2,5-Dichlorophenol