Order Entry
Switzerland
ContactUsLinkComponent
 
Organic reference standard, 3,5-Dinitrotoluene 100 µg/ml in methanol
  PROONE6513
 :  
Organic reference standard, 3,5-Dinitrotoluene 100 µg/ml in methanol
  PROONE6513
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    4,5 mL
  • CAS No.:
    618-85-9
  • Description:
    3,5-Dinitrotoluene 100 µg/ml in methanol
  • Formula:
    C₇H₆N₂O₄
  • Description:
    3,5-Dinitrotoluene