Organic reference standard, 2,4-Dinitrotoluene 100 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERXA12786200AL
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:121-14-2
- Description:2,4-Dinitrotoluene 100 µg/ml in acetonitrile
- Formula:C₇H₆N₂O₄
- Description:2,4-Dinitrotoluene