3-Phenyl-2-propyn-1-ol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1504-58-1
:
Formula:C₉H₈O
MDL Number:MFCD00040914
:132.16 g/mol
Boiling Pt:129…130 °C (10 mmHg)
Density:1.078
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
Related Information
Frequently Bought Together