Order Entry
Switzerland
ContactUsLinkComponent
3-Phenyl-2-propyn-1-ol 98%
Irritant
3-Phenyl-2-propyn-1-ol 98%
Certificates
undefined
Irritant
3-Phenyl-2-propyn-1-ol 98%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5830 to 1.5870 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together