Order Entry
Switzerland
ContactUsLinkComponent
Jars, with screw cap
Jars, with screw cap
Catalog # BRND62310
Supplier:  Brand
undefined
Jars, with screw cap
Catalog # BRND62310
Supplier:  Brand

Specifications

  • Ext.Ø:
    57 mm
  • Capacity:
    30 ml
  • Height:
    32 mm
  • Pk:
    500

Specifications

About this item

PP, translucent.

  • Conical shape
  • Excellent chemical resistance
  • Excellent temperature resistance

Ideal for sampling, transportation and storage.