Order Entry
Switzerland
ContactUsLinkComponent
Acetonitrile, ultra-low water content ≥99.5% (by GC, corrected for water content), BakerDRY™, J.T.Baker®
Flammable
Irritant
Acetonitrile, ultra-low water content ≥99.5% (by GC, corrected for water content), BakerDRY™, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Acetonitrile, ultra-low water content ≥99.5% (by GC, corrected for water content), BakerDRY™, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
  • ACS - Assay (CH₃CN) (by GC, corrected for water):
    ≥99.5 %
  • Appearance:
    Passes Test
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤1 ppm
  • ACS - Titrable Acid (µeq/g):
    ≤0.8
  • ACS - Titrable Base (µeq/g):
    ≤0.6
  • ACS - Water (by KF, coulometric):
    ≤10.0 ppm

Specification Test Results

Frequently Bought Together