Stopcocks, Needle-Type, Nalgene®
NALGENE® tanks up to 100 gals., 1-1/8 in.-12 straight female thread
Catalog # NALG6421-0010
Supplier: Thermo Fisher Scientific
Stopcocks, Needle-Type, Nalgene®
Catalog # NALG6421-0010
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PP, white.
- Excellent chemical resistance
- Excellent temperature resistance
Ensure safe and efficient liquid dispensing from tanks by using Nalgene™ spigots for storage tanks. Spigots are for use with Nalgene™ tanks up to 100 gal. with factory installed threaded boss.
Specifications
- For:NALGENE® tanks up to 100 gals.
- Thread:1-1/8 in.-12 straight female thread
- Cat. No.:NALG6421-0010
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact