Acetaldehyde for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-07-0
:
Formula:CH₃CHO
MDL Number:MFCD00006991
:44.05 g/mol
Merck Index:12,00037
Boiling Pt:20.1 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.78 g/cm³ (20 °C)
EINECS:200-836-8
UN:1089
ADR:3,I
:M-Clarity™ quality level = MQ200
Flash Pt:–27 °C
:GHS02
:GHS08
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together