Caprinaldehyde
Glass bottle, 100 mL
Catalog # ROTH7547.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Decylaldehyde, Decanal, Decyl aldehyde
- CAS Number:112-31-2
- Formula:C₁₀H₂₀O
- MDL Number:MFCD00007031
- Molecular Weight:156.27 g/mol
- Boiling Pt:207 °C (1013 hPa)
- Melting Pt:7 °C
- Density:0,83 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:203-957-4
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Cat. No.:ROTH7547.1



