Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Caprinaldehyde 95%

Caprinaldehyde 95%

Irritant
Caprinaldehyde 95%
Certificates
Caprinaldehyde 95%

2 Options Available Below

Product Details & Documents
Synonyms:  Decylaldehyde, Decanal, Decyl aldehyde
  • CAS Number:112-31-2
  • Formula:C₁₀H₂₀O
  • MDL Number:MFCD00007031
  • Molecular Weight:156.27 g/mol
  • Boiling Pt:207…209 °C (760 mmHg)
  • Melting Pt:7 °C
  • Density:0.83 g/cm³
  • Flash Pt:85 °C
  • Storage Temperature:Ambient
  • EINECS:203-957-4

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4270 to 1.4320 (20°C, 589 nm)
  • GC:
    ≥94 %
  • NMR (1H-NMR):
    Consistent with structure
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 34.70
    SFr. 34.70
    1 * 100 mL
       SFr. 34.70

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 88.20
    SFr. 88.20
    1 * 500 mL
       SFr. 88.20

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.