Silica gel drying agent, with moisture indicator (orange gel)
Supplier: Roth Carl
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Storage Temperature:Ambient
Frequently Bought Together