Order Entry
Switzerland
ContactUsLinkComponent
 
Tin(II) chloride dihydrate ≥98%, AnalaR NORMAPUR® analytical reagent (max. 0.05 ppm Hg)
  2512.0250
 :  
undefined
Tin(II) chloride dihydrate ≥98%, AnalaR NORMAPUR® analytical reagent (max. 0.05 ppm Hg)
  2512.0250
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Assay:
    Min. 98 %
  • Insolubility in diluted HCl:
    Passes test
  • NH4 (Ammonium):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • As (Arsenic):
    Max. 1 ppm
  • Ca (Calcium):
    Max. 0,005%
  • Cu (Copper):
    Max. 10 ppm
  • Fe (Iron):
    Max. 20 ppm
  • Hg (Mercury):
    Max. 0.05 ppm
  • K (Potassium):
    Max. 50 ppm
  • Na (Sodium):
    Max. 100 ppm
  • Pb (Lead):
    Max. 50 ppm

 

 

 :
 :225.65 g/mol
 :13,08861
 :623 °C (1013 hPa)
 :246 °C
 :2.71 g/cm³ (25 °C)
 :Ambient
 :231-868-0
 :3260
 :8,III
 :GHS07
 :GHS05
 :10025-69-1
 :SnCl₂·2H₂O
 :MFCD00149863