Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Tin(II) chloride dihydrate 98.0-103.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
 
undefined
 
 
Tin(II) chloride dihydrate 98.0-103.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
 

 

  • Assay:
    98.0 - 103.0 %
  • Identification:
    Passes test
  • Insolubility in diluted HCl:
    Passes test
  • NH4 (Ammonium):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • As (Arsenic):
    Max. 1 ppm
  • Ca (Calcium):
    Max. 0,005%
  • Cu (Copper):
    Max. 10 ppm
  • Fe (Iron):
    Max. 20 ppm
  • K (Potassium):
    Max. 50 ppm
  • Na (Sodium):
    Max. 100 ppm
  • Pb (Lead):
    Max. 50 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

 

Frequently Bought Together