Tin(II) chloride dihydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:10025-69-1
:
Formula:SnCl₂·2H₂O
MDL Number:MFCD00149863
:225.65 g/mol
Merck Index:13,08861
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:2.71 g/cm³ (25 °C)
UN:3260
ADR:8,II
:M-Clarity™ quality level = MQ300
EINECS:231-868-0
:GHS05
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:1187 g/l (20 °C)
- Melting Point:246 °C (anhydrous substance)
- Molar Mass:225.63 g/mol
- Bulk Density:1250 kg/m3
- Boiling Point:623 °C (1013 hPa) (anhydrous substance)
- Density:2.71 g/cm3 (20 °C)
- pH:1 - 2 (100 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.07815.0100_TDS_EN ProductPage_107815
Product specification
msds_107815
SDS
Frequently Bought Together