Order Entry
Switzerland
ContactUsLinkComponent
Magnesium nitrate hexahydrate 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
Oxidizing
Magnesium nitrate hexahydrate 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Oxidizing
Magnesium nitrate hexahydrate 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • pH (25°C; 5 %):
    5.0 - 7.0
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • NH4 (Ammonium):
    Max. 30 ppm
  • PO4 (Phosphate):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Ba (Barium):
    Max. 50 ppm
  • Ca (Calcium):
    Max. 100 ppm
  • Fe (Iron):
    Max. 5 ppm
  • K (Potassium):
    Max. 10 ppm
  • Mn (Manganese):
    Max. 5 ppm
  • Na (Sodium):
    Max. 50 ppm
  • Sr (Strontium):
    Max. 50 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Related Information

Frequently Bought Together