Strontium chloride hexahydrate, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:10025-70-4
:
Formula:SrCl₂·6H₂O
MDL Number:MFCD00149865
:266.62 g/mol
Merck Index:13,08920
Boiling Pt:100 °C (1013 hPa)
Melting Pt:61 °C
Density:1.93 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:233-971-6
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:2058 g/l (40 °C)
- Melting Point:61 °C
- Molar Mass:266.62 g/mol
- Bulk Density:1050 kg/m3
- Boiling Point:100 °C
- Density:1.95 g/cm3 (20 °C)
- pH:5 - 7 (50 g/l, H2O, 25 °C)
Specification Test Results
Frequently Bought Together