Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
About this item
Synonyms: Magnesium dichloride
:
:203.3 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:117 °C
Density:1.57 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7791-18-6
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781