Order Entry
Switzerland
ContactUsLinkComponent
Sodium tungstate dihydrate 99+% for analysis
Sodium tungstate dihydrate 99+% for analysis
Certificates
undefined
Sodium tungstate dihydrate 99+% for analysis

Specification Test Results

  • Appearance:
    White Powder or flakes
  • Assay:
    ≥99.0 %
  • Chloride (Cl):
    ≤50 ppm
  • Sulfate (SO4):
    ≤100 ppm
  • Arsenic (As):
    ≤5 ppm
  • Molybdenum (Mo):
    ≤20 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Free alkali (NaOH):
    ≤0.05 %

Specification Test Results

Related Information

Frequently Bought Together