Order Entry
Switzerland
ContactUsLinkComponent
Sodium sulphate decahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Sodium sulphate decahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Sodium sulphate decahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    420 g/l (20 °C)
  • Melting Point:
    32.4 °C
  • Molar Mass:
    322.19 g/mol
  • Bulk Density:
    700 kg/m3
  • Density:
    1.464 g/cm3
  • pH:
    5 - 7.5 (100 g/l, H2O, 20 °C) (anhydrous substance)

Specification Test Results

Related Information

Frequently Bought Together