Order Entry
Switzerland
ContactUsLinkComponent
Lithium sulphate monohydrate, dried ≥99.0% (dried basis, T), Puriss. p.a. ACS, Fluka™
Irritant
Lithium sulphate monohydrate, dried ≥99.0% (dried basis, T), Puriss. p.a. ACS, Fluka™
Certificates
Irritant
Lithium sulphate monohydrate, dried ≥99.0% (dried basis, T), Puriss. p.a. ACS, Fluka™

Specification Test Results

  • Assay (excluding dried substance):
    99.0 - 101.0 %
  • Water insoluble matter:
    Max. 0.01 %
  • Loss on drying (150°C):
    13.0 - 15.0 %
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 20 ppm
  • Nitrate (NO₃):
    Max. 10 ppm

Specification Test Results

Frequently Bought Together