
Lithium sulphate monohydrate 99+%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10102-25-7
:
Formula:Li₂SO₄·H₂O
MDL Number:MFCD00149766
:127.96 g/mol
Melting Pt:860 °C
Density:2,05 g/cm³ (20 °C)
EINECS:233-820-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Titration after Ion exchange:≥99.0 % (On dry substance)
- Loss on ignition:13.5 % to 15.0 % (600°C, 2 h)
Specification Test Results
Frequently Bought Together